Learn More
GSC Go Science Crazy Wire Gauze
Aids in the distribution of heat while using a Bunsen burner. GSC International Wire Gauze features rolled smooth wire edges. A variety of sizes are available.
Supplier: GSC Go Science Crazy 4000512
Alert:
Intended for ages 13 and up.
Description
- Wire gauze is used with Bunsen burners to aid in the distribution of heat
- Helps protect lab equipment
- Features rolled smooth wire edges
- Available in several sizes
Specifications
Wire Gauze | |
5 in. | |
Rolled smooth wire edges | |
5 x 5 in. |
Distribution of Heat with Bunsen Burners | |
5 in. | |
Wire |