Learn More
Grainger Wire Rope Locking Stud RH Thread 3/16 in
Supplier:  Grainger 16X725
Encompass_Preferred
The Grainger Approved Wire Rope Locking Stud is a 3/8" diameter wire rope stud with a working load limit of 1,260 lbs. It is made from drop forged carbon steel and zinc plated for corrosion resistance. The stud has a threaded stud on one end to accept a nut and a socket on the other end to accept a swaged wire rope fitting. It can be used for making wire rope connections in lifting applications. The stud meets ASME B30.26 specifications.
Catalog No. 50-276-8016