Learn More
Grainger Wire Rope Oval Sleeve 1/16 in Copper
Supplier:  Grainger 16X803
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope end fitting made of drop forged steel with a zinc finish. It is designed to terminate wire rope by inserting the rope and compressing with a press. This oval sleeve accommodates 1/4" wire rope, has an opening width of 0.28", fits rope diameters from 0.20" to 0.25", and weighs 0.06 lbs. The working load limit is 1,260 lbs. This wire rope end fitting provides a permanent loop and is ideal for creating attachments and connections.
Catalog No. 50-276-7970
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.