Learn More
Grainger Wire Rope Oval Sleeve 1/8 in Copper
Supplier:  Grainger 16X778
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of drop forged steel. It is designed to fit 1/4" wire rope and has an opening width of 0.28". The working load limit is 1,260 lbs. This wire rope sleeve can be used for a variety of rigging and load securement applications. Its drop forged steel construction provides durability. With its 1/4" wire rope compatibility and 1,260 lb working load limit, this LOOS sleeve is suitable for light to medium duty wire rope rigging needs.
Catalog No. 50-276-7223
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.