Learn More
Grainger Wire Rope Oval Sleeve 1/8 in Copper
Supplier:  Grainger 16X790
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of galvanized steel. It has an inside diameter of 1/2", fits 1/8" wire rope, and is 7/8" long. This wire rope sleeve is designed to protect the ends of wire rope from fraying and unraveling. It can be used in a variety of wire rope termination applications. The galvanized coating provides corrosion resistance. This wire rope sleeve meets military specification MIL-S-781. It is sold individually.
Catalog No. 50-276-7906
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.