Learn More
Grainger Wire Rope Oval Sleeve 3/16 in Copper
Supplier:  Grainger 16X779
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of malleable cast iron. It has an inside diameter of 1-1/4", fits 1/4" wire rope, and is 7/8" long. This wire rope sleeve is uncoated and is rated at a safe working load of 1 ton. It can be used to form eyes on the ends of wire rope for attachments. This wire rope oval sleeve is made in the USA by Loos & Co.
Catalog No. 50-276-7225
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.