Learn More
Grainger Wire Rope Oval Sleeve 7/16 in Copper
Supplier:  Grainger 16X812
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of galvanized steel. It has an inside diameter of 16mm and is used to form an eye at the end of a wire rope for attaching fittings. The working load limit is 1,300 lbs and the break strength is 2,600 lbs. The sleeve is 13mm wide x 36mm long. To install, insert rope and swage sleeve with the proper swaging tool to recommended torque. It can be used with 6x7 and 6x19 wire rope. This wire rope end fitting provides a secured loop for safe load attachment.
Catalog No. 50-276-7971
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.