Learn More
Grainger Wire Rope Stop Sleeve 1/4 in Copper
Supplier:  Grainger 16X822
Encompass_Preferred
The Loos Wire Rope Stop Sleeve Stop is a wire rope termination device designed to prevent fraying of wire rope ends. It is made from drop-forged steel and zinc plated for corrosion resistance. This wire rope stop can be used on 6x7 and 6x19 wire rope constructions from 1/8" to 3/4" diameter. It installs by sliding over the bitter end of the wire rope and compressing with a hammer to grip the rope strands. This helps prevent unraveling and provides a finished look to exposed wire rope ends in rigging applications. The working load limit is not specified.
Catalog No. 50-276-7806
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.