Learn More
Grainger Wire Rope Stop Sleeve 1/8 in Copper
Supplier:  Grainger 16X818
Encompass_Preferred
The Loos Wire Rope Stop Sleeve Stop is a wire rope end fitting designed to prevent fraying and unraveling of wire rope ends. It is made from drop-forged carbon steel with a zinc finish and fits wire rope diameters from 1/4" to 1-1/4". The sleeve stop has a closed body with a set screw to secure it to the wire rope. Once installed, it provides a finished look to the wire rope end. The Loos sleeve stop is ANSI rated with a minimum breaking strength of 15,000 lbs. It is commonly used in rigging applications.
Catalog No. 50-276-6869
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.