Learn More
Grainger Wire Rope Thimble 5/16in Rope Dia Steel
Supplier:  Grainger 16X765
Encompass_Preferred
The Loos Wire Rope Thimble is a galvanized steel thimble with a 3/8" inside diameter. It is compatible with 3/8" wire rope and is used to prevent fraying or damage to the rope ends. The thimble has a rounded rim to prevent chafing and has a standard weight pattern. It meets military specification MIL-T-781J and is made in the USA by Loos & Co. This thimble provides a secure loop termination for lifting, pulling, and tensioning applications.
Catalog No. 50-276-7905
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.