Learn More
Grainger Wire Rope Thimble 5/32 in Rope Dia SS
Supplier:  Grainger 16X769
Encompass_Preferred
The Loos Wire Rope Thimble is made of galvanized steel with a 5/32 inch inside diameter. It is designed to fit 5/32 inch wire rope and protect it from wear. The thimble has a closed round eye and tapered barrel. It can be used in a variety of rigging applications to prevent fraying and extend the life of wire rope. The thimble is rated up to 1 ton working load limit.
Catalog No. 50-276-5117
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.