Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Wire Stripper
SDPSCI_ED

Catalog No. S99397 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (English):
4 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S99397 4 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S99397 Supplier United Scientific™ Supplier No. WSTK01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Wire Stripper quickly removes insulation without damaging wire.

  • Set-screw adjusts stripper to fit wire gauges from 10 to 24.
  • Made from drop-forged, heat-treated steel with black baked enamel finish.
  • PVC handles.
  • Total length: 4 in. (10.1 cm)

Specifications

Blade Material Heat-treated Steel
Product Type Wire Stripper
For Use With (Equipment) Wire gauges from 10 to 24
Grade Standard
Housing Material PVC Handle
Length (English) 4 in.
Length (Metric) 10.1 cm
Material Steel
For Use With (Application) To quickly removes insulation without damaging wire
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.