Learn More
Thermco Products Wireless Temperature Probes Email / SMS Alerting VFC Data Logger
Catalog No. 04500041
Change view
Click to view available options
For Use With (Equipment):
Data Loggers
Catalog No. For Use With (Equipment)
04-500-041 Data Loggers
1 options
Catalog No. 04-500-041 Supplier Thermco Products Supplier No. ACCSL2021
Add to Cart
Add to Cart
Kit includes only 1 wireless sensor, additional sensor listed below, 1 Wireless Vaccine Bottle Sensors with 2 Years NIST Traceable Certificate, USB Cable, 110 V USB Power Adapter, 3 x AA batteries, 2 x AAA batteries.
  • The AccuTherm VFC SmartLOG 2021 Data Logger with wireless temperature sensors monitors vaccines up to 30 ft. away, eliminating the need for wires through refrigerator or freezer doors.
  • Serialized, NIST-traceable wireless sensors with CDC-required glycol buffer bottles can be placed inside storage units to track real-time temperatures.
  • Color touchscreen display shows current, minimum, and maximum temperatures, sensor signal strength, battery status, and ambient room conditions.
  • WiFi connectivity enables remote access through a browser-based dashboard on PCs or smartphones, with no cloud subscription required.
  • SMS and email alerts notify users of temperature excursions, power loss, or signal issues, with additional notifications when conditions return to normal.
  • Optional daily status reports at 1 am and 1 pm provide temperature and power updates.
  • Internal 2 MB memory supports up to 2 Years of data storage at 5-minute logging intervals.
  • Generates PDF reports directly without the need for additional software.
  • Hot-swappable sensors eliminate recalibration downtime and ensure uninterrupted monitoring.
  • Meets CDC and VFC standards for vaccine temperature monitoring.
Specifications
Type Probe
For Use With (Equipment) Data Loggers