Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Wiskostatin
GREENER_CHOICE

Catalog No. p-4816630
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-341-0 10 mg
44-345-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-341-0 Supplier Tocris Bioscience™ Supplier No. 4434/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

N-WASP inhibitor; inhibits Arp2/3 activation

Inhibitor of neural Wiskott-Aldrich syndrome protein (N-WASP) activity. Interacts with the regulatory GTPase-binding domain of N-WASP; inhibits activation of Arp2/3 complex by maintaining N-WASP in an inactive conformation. Also inhibits PIP2-induced actin polymerization (EC50 ∽ 4μM).

Chemical Identifiers

CAS 253449-04-6
Molecular Formula C17H18Br2N2O

Specifications

Quantity 10 mg
Formula Weight 426.15
Purity >99%
Molecular Weight (g/mol) 426.15
Chemical Name or Material Wiskostatin
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.