Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard™ Magnetic 96 DNA Plant System

Catalog No. PRFF3761 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Product Type:
96 DNA Plant System
Replacement Wash Buffer
Quantity:
4 x 96 Preps
40 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
PRFF3761 96 DNA Plant System 4 x 96 Preps
PRA3811 Replacement Wash Buffer 40 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRFF3761 Supplier Promega Supplier No. FF3761
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides manual or automated 96-well, high-throughput purification of DNA from plant leaf and seed tissue.

  • Automated Purification of DNA from Leaves and Seeds
  • Cuts purification time compared to CTAB extraction
  • Eliminates organic extractions.
  • Validated for both leaf and seed tissue by PCR and RAPD analysis

Specifications

Content And Storage 15°C to 30°C
Format Kit
Isolation Technology Magnetic Bead
No. of Reactions 4 x 96 Preps
Product Type 96 DNA Plant System
Quantity 4 x 96 Preps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.