Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard™ Magnetic DNA Purification System for Food

Catalog No. PRFF3751 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
No. of Reactions:
200 Preps
400 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
PRFF3751 400 Preps
PRFF3750 200 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRFF3751 Supplier Promega Supplier No. FF3751
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

MagneSil Paramagnetic Particles; Lysis Buffer A, Food; Lysis Buffer B, Food; Precipitation Solution, Food; RNase A Solution

Requires

MagneSphere Technology Magnetic Separation Stand (Z5332 or Z5342)

The Wizard™ Magnetic DNA Purification System for Food is designed for purification of DNA from a variety of food samples.

  • Rapid Isolation of DNA from Many Food Types
  • Requires minimal centrifugation and eliminates organic extractions
  • Purified DNA suitable for PCR-based GMO testing

U.S. Patents 6,027,945 and 6,368,800, Australian Patent 732756 and other patents and patents pending. no value

Specifications

Content And Storage 15°C to 30°C
Description Magnetic DNA Purification System for Food
Format Kit
Isolation Technology Magnetic Bead
No. of Reactions 400 Preps
No. of Samples 400
Product Type System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.