Learn More
Description
- More than 1mg Plasmid from 1L Cultures
- 3-hour protocol
- Resin-based batch column method
- No organic extractions
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Quantity | 1 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.