Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard™ SV 96 Cell Lysis Solution

Catalog No. PRA7128 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
800 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA7128 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA7128 Supplier Promega Supplier No. A7128
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell Lysis Solution is a component of the Wizard™ MagneSil, Wizard™ Plus and Wizard™ SV 96 and SV 9600 Plasmid DNA Purification Systems and the PureYield Plasmid Midiprep System.

  • Reagent Used in Plasmid DNA Isolation Protocols
  • Cat.# A7122 is a component of the Wizard(R) Plus DNA Purification Systems
  • Cat.# A7125 is a component of the PureYield™ Plasmid Midiprep System
  • Cat.# A7123 and A7128 are components of the Wizard(R) SV 96 and SV 9600 Plasmid DNA Purification Systems
  • Cat.# A7124 is a component of the Wizard(R) MagneSil(R) Plasmid Purification System

Specifications

Content And Storage 15°C to 30°C
Quantity 800 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.