Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard™ SV 96 Cell Resuspension Solution

Catalog No. PRA7118 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
800 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA7118 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA7118 Supplier Promega Supplier No. A7118
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Cell Resuspension Solution is a component of the Wizard™ MagneSil, Wizard™ Plus and Wizard™ SV 96 Plasmid DNA Purification Systems and the PureYield Plasmid Midiprep System.

  • Reagent Used in Plasmid DNA Purification Protocols
  • Cat.# A7112 is a component of the Wizard(R) Plus DNA Purification Systems
  • Cat.# A7115 is a component of the PureYield™ Plasmid Midiprep System
  • Cat.# A7113 and A7118 are components of the Wizard(R) SV 96 Plasmid DNA Purification System
  • Cat.# A7114 is a component of the Wizard(R) MagneSil(R) Plasmid Purification System

Specifications

Content And Storage 15°C to 30°C
Quantity 800 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.