Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard SV 96 Lysate Clearing Plates, 100 pack

Catalog No. PRA2248 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Array:
8 x 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Array
PRA2248 8 x 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA2248 Supplier Promega Supplier No. A2248
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Wizard SV 96 Lysate Clearing Plates are used with the Wizard SV 96 Binding Plates and Vac-Man 96 Vacuum Manifold for simultaneous lysate clearing and DNA binding in the Wizard SV 96 and SV 9600 Plasmid Purification System protocols.

The Wizard™ SV 96 Lysate Clearing Plates are designed for use in the Wizard™ SV 96 and SV 9600 Plasmid Purification System protocols.

  • Function: Used for simultaneous lysate clearing and DNA binding.
  • Compatibility: Designed for use with Wizard™ SV 96 Binding Plates (Cat.# A2271 and A2278) and Vac-Man™ 96 Vacuum Manifold (Cat.# A2291).
  • Use Restrictions: For Research Use Only. Not for Use in Diagnostic Procedures.

Specifications

Format Standard
Content And Storage 15°C to 30°C
Array 8 x 12
Product Type Lysate Clearing Plate
No. of Wells 96
Quantity 100 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.