Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Wizard™ SV 96 Neutralization Solution

Catalog No. PRA1488 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
950 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA1488 950 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA1488 Supplier Promega Supplier No. A1488
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Neutralization Solution is a component of the Wizard™ MagneSil, Wizard™ MagneSil™ Tfx, Wizard™ Plus and Wizard™ SV 96 Plasmid DNA Purification Systems and the PureYield Plasmid Midiprep System.

  • Reagents Used in DNA Plasmid DNA Purification Protocols
  • Cat.# A7131 is a component of the Wizard MagneSil Tfx™ and Wizard(R) Plus DNA Purification Systems
  • Cat.# A7132 is a component of the Wizard(R) MagneSil(R) Plasmid Purification System
  • Cat.# A1485 is a component of the PureYield™ Plasmid Midiprep System
  • Cat.# A1488 is a component of the Wizard(R) SV 96 Plasmid DNA Purification System

Specifications

Content And Storage 15°C to 30°C
Format Bottle
Product Type Neutralization Solution
Quantity 950 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.