Learn More
Eurofins Discoverx WKYMVm-NH2, 1 mg. (92-1075)
Supplier:  Eurofins Discoverx 921075
Encompass
WKYMVm-NH2 is a peptide found in immune cells that is an agonist of formyl peptide receptors (FPR), with EC50 of 75 pM, 75 pM, and 3 nM for receptors 1, 2, and 3, respectively. It induces calcium mobilization, superoxide production, and chemotaxic migration in monocytes and neutrophils. Assists in neutrophil survival and stimulates phospholipase C (PLC) mediated InoP formation. Provided as 1 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9615
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.