Learn More
TARGETMOL CHEMICALS INC WM-3835 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T910210MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. WM-3835 is a novel and high-specific small molecule inhibitor of Lysine Acetyltransferase 7 (KAT7 MYST2 HBO1) able to potently suppressed OS cell proliferation and migration and leads to apoptosis activation. purity: 97%
Catalog No. 50-257-6212
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.