Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wnt-9a Antibody (326707) [Janelia Fluor™ 525], Novus Biologicals™
SDP

Catalog No. NB652640
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB652640 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB652640 Supplier R&D Systems Supplier No. FAB31571JF525

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Wnt-9a Monoclonal antibody specifically detects Wnt-9a in Human samples. It is validated for Immunohistochemistry,Western Blot

Specifications

Antigen Wnt-9a
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 326707
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias MGC138165, MGC141991, Protein Wnt-14, wingless-type MMTV integration site family, member 14, wingless-type MMTV integration site family, member 9A, WNT14protein Wnt-9a
Host Species Mouse
Immunogen E. coli-derived recombinant human Wnt-9a, Pro48-Glu92 & Arg231-Phe304, Accession # O14904.2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7483
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.