Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Wnt-9b Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3669SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3669SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3669SP Supplier R&D Systems Supplier No. AF3669SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

Wnt-9b Polyclonal specifically detects Wnt-9b in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Wnt-9b
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O35468
Gene Alias Protein Wnt-14b, wingless-type MMTV integration site family, member 15, wingless-type MMTV integration site family, member 9B, Wnt-14b, WNT14Bprotein Wnt-9b, Wnt-15, WNT15Protein Wnt-15, Wnt9b
Gene Symbols WNT9B
Host Species Goat
Immunogen E. coli-derived recombinant mouse Wnt-9b Ala46-Glu90, Lys228-Tyr298 Accession # O35468
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7484
Test Specificity Detects mouse Wnt-9b in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) Wnt-2, rhWnt-9a, recombinant mouse (rm) Wnt-1, rmWnt-2b, rmWnt-3a, rmWnt-4, rmWnt-5a, rmWnt-5b, rmWnt-11, and rmWnt-16 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.