Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNT10B Monoclonal Antibody (5A7)
GREENER_CHOICE

Catalog No. PIMA515557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515557 Supplier Invitrogen™ Supplier No. MA515557
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15557 targets WNT10B in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15557 immunogen is purified recombinant fragment of human WNT10B expressed in E. Coli. MA5-15557 detects WNT10B which has a predicted molecular weight of approximately 43kDa.

The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It may be involved in breast cancer, and its protein signaling is likely a molecular switch that governs adipogenesis. This protein is 96% identical to the mouse Wnt10b protein at the amino acid level. This gene is clustered with another family member, WNT1, in the chromosome 12q13 region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT10B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5A7
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene WNT10B
Gene Accession No. O00744
Gene Alias Protein Wnt-10b; Protein Wnt-12; SHFM6; wingless related MMTV integration site 10b; wingless-type MMTV integration site family member 10B; wingless-type MMTV integration site family, member 10B; Wnt family member 10B; Wnt10; WNT10B; Wnt-10b; WNT-10B protein; WNT12; wnt-12
Gene Symbols WNT10B
Host Species Mouse
Immunogen Purified recombinant fragment of human WNT10B expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7480
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.