Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WNT5A Monoclonal Antibody (6F2)
GREENER_CHOICE

Catalog No. PIMA515502 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515502 Supplier Invitrogen™ Supplier No. MA515502
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15502 targets WNT5A in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15502 immunogen is purified recombinant fragment of WNT5A expressed in E. Coli.. MA5-15502 detects WNT5A which has a predicted molecular weight of approximately 42.3kDa.

Wnt5A is a secreted protein found in the extracellular region that belongs to the Wnt family. It is a proto-oncogene that functions in modulating cell fate and behavior during vertebrate development and has an important role in Wnt signaling pathway. It is a glycoprotein that plays an important role in cell communication and development. Wnt5A acts as a ligand for members of the frizzled family of seven transmembrane receptors. It is suspected that Wnt5A is a developmental protein. Wnt5A is a signaling molecule which affects the development of discrete regions of tissues and is likely to signal over only few cell diameters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT5A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6F2
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene WNT5A
Gene Accession No. P41221
Gene Alias 8030457G12Rik; hWNT5A; OTTHUMP00000212837; OTTHUMP00000212861; Protein Wnt-5a; wingless-related MMTV integration site 5A; wingless-type MMTV integration site 5A; wingless-type MMTV integration site family member 5A; wingless-type MMTV integration site family, member 5A; Wnt family member 5A; WNT5A; Wnt-5a; WNT-5A protein
Gene Symbols WNT5A
Host Species Mouse
Immunogen Purified recombinant fragment of WNT5A expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7474
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.