Learn More
Invitrogen™ WNT6 Polyclonal Antibody

Description
The antibody detects endogenous levels of total WNT6 protein.
Specifications
Specifications
| Antigen | WNT6 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | WNT6 |
| Gene Accession No. | P22727, Q9Y6F9 |
| Gene Alias | AA409270; Protein Wnt-6; wingless-related MMTV integration site 6; wingless-related MMTV integration site 6 homolog; wingless-type MMTV integration site family member 6; wingless-type MMTV integration site family, member 6; Wnt family member 6; WNT6; Wnt-6 |
| Gene Symbols | WNT6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.