Learn More
Apexbio Technology LLC Wogonin 632-85-9 20mg
SDP
Supplier:  Apexbio Technology LLC N128320
Encompass_Preferred
Wogonin (CAS 632-85-9) is a bioactive flavonoid that acts as a selective inhibitor of cyclooxygenase-2 (COX-2) with an IC50 value of 46 M and does not affect cyclooxygenase-1 (COX-1) activity It also suppresses the induction of inducible nitric oxide synthase (iNOS) thereby mitigating nitric oxide-mediated inflammatory pathways In cellular studies wogonin attenuates COX-2 and iNOS expression reducing activation-induced cell death in C6 glial cells These properties position wogonin as a valuable tool for investigating inflammation signaling and neuroinflammatory processes in biomedical research
Catalog No. 50-000-97093
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.