Learn More
Bulwark Women's FR/CP Lab Coat
Catalog No. 19999545 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-999-545 2X-Large
19-999-540 X-Small
19-999-541 Small
19-999-542 Medium
19-999-543 Large
19-999-544 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-999-545 Supplier Bulwark Supplier No. KNR7RB 2L 00
Add to Cart
Add to Cart
Flame resistant/chemical protective lab coat brings together the latest innovations in lab safety to protect wearers against multiple hazards.
  • Lightweight and breathable fire resistant/chemical protective lab coat.
  • Black lay-down collar for easy recognition as a CP lab coat.
  • Splash resistant pass-through for inner pocket access.
  • Flame-resistant 4.5 oz. Nomex IIIA fabric prevents clothing ignition during short-duration thermal incidents and emergency exposure to flame.
  • Repels small amounts of liquid chemicals that are either dropped or splashed on the fabric, resisting penetration and wicking of an inadvertent, secondary liquid chemical splash.
  • Concealed snap front neck closures.
  • One double-stitched chest pocket and two double-stitched lower pockets with flaps
  • Arc Rating ATPV 5.6 cal/cm2
  • Category 1 Protection
  • NFPA 2112 Compliant
Specifications
Product Type Chemical Resistant Lab Coat
Size 2X-Large
Gender Women's
Material Aramid Fiber
Color Royal Blue
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Flame, Chemicals
Style Regular
No. of Pockets 3
Collar Type Lay-down
Certifications/Compliance Category 1, NFPA 2112