Learn More
United Scientific Supplies Women's Laboratory Coats
SDPSCI_ED
Catalog No. S139587 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S139587 Large
S139585 Small
S139586 Medium
S139588 X-Large
S139589 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. S139587 Supplier United Scientific Supplies Supplier No. LCLL01
Add to Cart
Add to Cart
This item is not returnable. View return policy
Allows for safety and comfort in the laboratory.
  • Knee-length lab coat made of easy-care, white cotton blend fabric (65% polyester, 35% cotton)
  • One upper left breast pocket, two roomy lower pockets, side pocket slit openings, and a sewn-down back belt
  • Tapered and cut for a better fit than unisex or men's lab coats
Specifications
Product Type General Purpose Lab Coat
Size Large
Gender Women's
Material Polyester, Cotton
Color White
Closure Type Buttons
Cuff Style Hemmed
Disposable Reusable
Sterility Non-sterile
Resistance Light Liquid Splash
Style Standard Length
No. of Pockets 2