Learn More
Description
- Flame resistant women's lab coat is designed for functionality, comfort, and safety
- CAT 1 arc rating, NFPA 2112 compliant
- Rib knit cuffs
- Concealed and reinforced snap front closures for easy donning and doffing
- Side vents at hem for ease of walking
- Four exterior pockets that provide ample storage
- Certifications Level 1: NFPA 2112, ASTM F1506
- Certifications Level 2: Category 1 Protection
- Hazard : Flash Fire, Arc Flash
- Wash Care : Industrial Laundry - Light Soil
- Fabric: Body: 4.5 oz. (150 g/m2) Nomex IIIA Flame Resistant Plain Weave
- Blend: Body: 93% Nomex Aramid / 5% Aramid Fibers / 2% Other Fibers
- Protection: ATPV Arc Rating: 4.4
Specifications
Specifications
| Product Type | Flame-resistant Lab Coat |
| Size | X-Small |
| Gender | Women's |
| Material | Nomex IIIA |
| Color | Royal Blue |
| Closure Type | Snaps |
| Cuff Style | Knit |
| Disposable | Reusable |
| Sterility | Non-sterile |
| Resistance | Flame |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.