Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Wood Chair - Desk Height

Catalog No. 14000501 Shop All Fisherbrand Products
Change view
Click to view available options
Foot Type:
Black Dual, Self-braking, Non-marring Casters
Black Non-marring Bell Glides
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Foot Type
14-000-501 Black Dual, Self-braking, Non-marring Casters
14-359-316 Black Non-marring Bell Glides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-000-501
Add to Cart
Add to Cart

Fisherbrand™ Wood Chairs are height adjustable for 31 in. work surfaces and feature contoured seat and manual adjustments. 7-ply furniture grade hardwood in natural finish provides a classic look and feel.

  • Adjustable from 14-19.5 inches from the top of the seat to the floor
  • Features ergonomic adjustments such as seat height
  • Manual back height and depth adjustments
  • Black, nylon-reinforced fiberglass base
  • 7-ply furniture grade hard wood with natural finish

Specifications

Product Type General Seating Chair
With Lumbar Support No
Foot Type Black Dual, Self-braking, Non-marring Casters
With Tilt No
With Casters Yes
With Backrest Yes
Footrest No
With Armrest No
Load Bearing Capacity (English) 300 lb.
Seating Height Desk Height
Load Bearing Capacity (Metric) 136 kg
Color Natural
Base Material Nylon Reinforced Fiberglass
Adjustable (Height) 14 to 19.5 in.
Adjustable Yes
Back Style Standard
Material Plywood
Seating Type Desk Height
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.