Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Public Seating™ Wood Science Lab Table with Whiteboard Top
SCI_ED

Catalog No. S83631
Change view
Click to view available options
Dimensions (D x W x H):
48 x 24 x 30 in.
48 x 24 x 36 in.
54 x 24 x 30 in.
54 x 24 x 36 in.
60 x 14 x 36 in.
60 x 24 x 30 in.
60 x 30 x 30 in.
60 x 30 x 36 in.
60 x 42 x 30 in.
60 x 42 x 36 in.
72 x 24 x 30 in.
72 x 24 x 36 in.
72 x 30 x 30 in.
72 x 30 x 36 in.
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H)
S83631 72 x 30 x 30 in.
S83626 48 x 24 x 30 in.
S83654 48 x 24 x 36 in.
S83627 54 x 24 x 30 in.
S83655 54 x 24 x 36 in.
S83628 60 x 24 x 30 in.
S83656 60 x 14 x 36 in.
S83629 72 x 24 x 30 in.
S83657 72 x 24 x 36 in.
S83630 60 x 30 x 30 in.
S83658 60 x 30 x 36 in.
S83659 72 x 30 x 36 in.
S83632 60 x 42 x 30 in.
S83660 60 x 42 x 36 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. S83631 Supplier National Public Seating™ Supplier No. SLT13072W
Add to Cart
Add to Cart

NPS Industrial, Fixed Height Whiteboard Top is an asset in any setting.

  • Whiteboard is made from 1.25 in. industrial grade particleboard with a high-pressure laminate top and 3 mm edge banding
  • Solid ash legs and apron
  • Rubber liquid protecting boots
  • Legs attach with metal on metal double bolt through assembly on 13-gauge steel corner braces
  • Adjustable glides for accurate leveling
  • 10 year warranty

Specifications

Product Type Lab Table
Top Style Whiteboard
Color White
For Use With (Application) Science Labs
Certifications/Compliance MAS2, ANSI BIFMA
Dimensions (D x W x H) 72 x 30 x 30 in.
Warranty 10 years
Material Ash Wood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.