Learn More
Eisco™ Wooden Friction Block with Hook
SCI_ED
Catalog No. S27646 Shop All Eisco Products
Change view
Click to view available options
Material:
Wood/Felt
Wood/Sandpaper
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S27646 Wood/Sandpaper
S27645 Wood/Felt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27646 Supplier Eisco™ Supplier No. PH0201605S
Only null left
Add to Cart
Add to Cart
Wooden friction blocks with hooks are useful for teaching the basics of force and motion in a simple, yet effective way. A friction block used in tandem with a spring scale is a powerful tool for teaching students about friction, kinetic and potential energy, acceleration, and force
  • Wooden friction block with c-hook attached at one end
  • Use with a spring scale to measure force and mass
  • Features 1 wooden side and 1 side covered with a unique material for comparative study
Specifications
Product Type Friction Block
Material Wood/Sandpaper
For Use With (Application) Measuring Force and Mass
For Use With (Equipment) Friction Track
Includes C-Hook