Learn More
Eisco™ Wooden Friction Board Apparatus
SCI_ED
Catalog No. S27664 Shop All Eisco Products
Change view
Click to view available options
Includes:
(2) Friction Sliders with Hooks
Catalog No. Includes
S27664 (2) Friction Sliders with Hooks
1 options
Catalog No. S27664 Supplier Eisco™ Supplier No. PH0336
Add to Cart
Add to Cart
Wooden Friction Board is ideal for investigating sliding friction
  • Consists of a wooden base and a smooth metal surface measuring
  • Two friction sliders with hooks are included, each with a different top surface
  • Slider Dimensions: 5.5 x 3 x 0.75 in.
Specifications
Product Type Friction Board Apparatus
Material Wood
For Use With (Application) Investigating Sliding Friction
Includes (2) Friction Sliders with Hooks