Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Wooden Lens Holder
SCI_ED

Catalog No. S27410 Shop All Eisco Products
Change view
Click to view available options
Material:
Wood
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S27410 Wood
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27410 Supplier Eisco™ Supplier No. PH0550A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

V-shaped wooden lens holder for use with lenses and mirrors while demonstrating the principles of light and color.

  • Accommodates lenses or mirrors up to 76 mm diameter
  • Holds convex or concave lenses and mirrors
  • Index mark engraved on each end of the base
  • Dimensions: 5 x 3.75 x 1.87 in.
  • Lenses and mirrors not included

Specifications

Material Wood
For Use With (Application) Demonstrating the Principles of Light and Color
Product Type Lens Holder
Shape V-shaped
Dimensions (L x W x H) 5 x 3.75 x 1.87 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.