Learn More
Specifications
Specifications
| Certifications/Compliance | UL recognized, CSA accepted, AGA approved, Halogen free (DIN VDE 0472/part 815) |
| Description | Abrasion, Chemical, Corrosion, Humidity, UV and Weather-Resistant |
| Adhesive Type | Acrylic, Permanent |
| Color | White |
| Finish | Gloss |
| Material | Polyester (B-423) |
| Product Type | Thermal Transfer Printer Label |
| For Use With (Application) | Asset tracking, Barcoding, Component ID, Equipment ID, General purpose ID, Rating plates |
| For Use With (Equipment) | BMP™71 Printer |
| Quantity | 5000/Roll |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.