Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ WorkHorse™ Raised Panel Label Overlaminates

Catalog No. 19109445 Shop All Brady Corporation Products
Change view
Click to view available options
Size (English):
1.06 x 0.49 in. (W x H)
1.2 x 1.5 in. (W x H)
1.2 x 1.9 in. (W x H)
1.7 x 0.59 in. (W x H)
1.8 x 1.8 in. (W x H)
2 x 1 in. (W x H)
2.4 x 2.4 in. (W x H)
3 x 0.75 in. (W x H)
3 x 1 in. (W x H)
3 x 2.5 in. (W x H)
3 x 3.5 in. (W x H)
4 x 1 in. (W x H)
For Use With (Application):
For Raised Panel Labels
For Round Raised Panel Labels
Round Raised Panel Labels, w/o Notch
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English) For Use With (Application)
19-109-445 3 x 2.5 in. (W x H) For Raised Panel Labels
19-109-435 1.2 x 1.5 in. (W x H) Round Raised Panel Labels, w/o Notch
19-109-436 1.2 x 1.9 in. (W x H) For Round Raised Panel Labels
19-109-434 1.2 x 1.5 in. (W x H) For Round Raised Panel Labels
19-109-438 2.4 x 2.4 in. (W x H) For Round Raised Panel Labels
19-109-437 1.8 x 1.8 in. (W x H) For Round Raised Panel Labels
19-109-440 1.7 x 0.59 in. (W x H) For Raised Panel Labels
19-109-439 1.06 x 0.49 in. (W x H) For Raised Panel Labels
19-109-441 2 x 1 in. (W x H) For Raised Panel Labels
19-109-443 3 x 0.75 in. (W x H) For Raised Panel Labels
19-109-442 4 x 1 in. (W x H) For Raised Panel Labels
19-109-444 3 x 1 in. (W x H) For Raised Panel Labels
19-109-446 3 x 3.5 in. (W x H) For Raised Panel Labels
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 19-109-445 Supplier Brady™ Supplier No. THTOL177966B1
Add to Cart
Add to Cart

Cover and protect thermal transfer printable raised panel labels. Brady™ WorkHorse™ Raised Panel Label Overlaminates are made from polyester (B-966B).

  • Made of clear polyester
  • Made for Brady thermal transfer printable raised panel labels

Specifications

Adhesive Type Acrylic, Permanent
Color Clear
Format Roll
Material Polyester (B-966B)
Product Type Raised panel label overlaminate
For Use With (Application) For Raised Panel Labels
Size (English) 3 x 2.5 in. (W x H)
Thickness (English) 0.0029 in.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.