Description
- Tear-tape design to keep average fall arrest forces below 900 lb./6kN
- Made of stainless steel
- Increase product life cycle when working in corrosive environments such as mining, food processing, water treatment and oil, gas, and pet
Specifications
Specifications
| Color | Yellow |
| Material | Stainless Steel |
| Product Type | Energy-absorbing lanyards |
| Leg Configuration | Single-Leg |
| Certifications/Compliance | ANSI 359.13, OSHA |
| Connector Types | Harness connection: 36C; Anchorage connection: 36C |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.