Learn More
MSA™ Workman™ Energy-Absorbing Lanyards
Catalog No. 19825022 Shop All MSA Products
Change view
Click to view available options
Connector Types:
Harness connection: 36C; Anchorage connection: (2)36C
Harness connection: 36C; Anchorage connection: 36C
Leg Configuration:
Single-Leg
Twin-Leg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types Leg Configuration
19-825-022 Harness connection: 36C; Anchorage connection: (2)36C Twin-Leg
19-825-021 Harness connection: 36C; Anchorage connection: 36C Single-Leg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-825-022 Supplier MSA™ Supplier No. 10113159
For working in confined spaces and at heights. MSA™ Workman™ Energy-Absorbing Lanyards limits force on body and anchor location during fall.
  • Tear-tape design to keep average fall arrest forces below 900 lb./6kN
  • Made of stainless steel
  • Increase product life cycle when working in corrosive environments such as mining, food processing, water treatment and oil, gas, and pet
Specifications
Material Stainless Steel
Color Yellow
Connector Types Harness connection: 36C; Anchorage connection: (2)36C
Leg Configuration Twin-Leg
Product Type Energy-absorbing lanyards
Certifications/Compliance ANSI 359.13, OSHA
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.