Learn More
MSA™ Workman™ FP Stryder™ Beam Grip
Catalog No. 19166583 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
14 - 23.5 in. beam
4 - 13.5 in. beam
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-166-583 14 - 23.5 in. beam
19-166-582 4 - 13.5 in. beam
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-166-583 Supplier MSA™ Supplier No. 10144432
Add to Cart
Add to Cart
Workman FP Stryder™ Beam Anchors quickly and easily attach to an I-beam flange without the need for any tools.
  • Compatible for use with MSA fall arrest and confined space entry/retrieval components
  • Attaches to I-beam flange - no tools required
Compliance
Meets applicable OSHA standards; ANSI™ Z359.1-2007 requirements.
Specifications
For Use With (Equipment) 14 - 23.5 in. beam
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.