Learn More
Workrite™ Flame-Resistant Lab Coats
Catalog No. 19082410 Shop All Red Kap Industries Products
Change view
Click to view available options
Size:
Small
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-082-410 Small
19-082-413 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-082-410 Supplier Workrite™ Supplier No. 352NX60RBSM 0R
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Lab coat with concealed snap front closure and seamed back for closure.
Lab coat with concealed snap front closure and seamed back for closure
  • Made of 6 oz. Nomex™ IIIA fabric
  • Four chevron pockets for ample storage
  • Side pass-thru for access to inner clothing
  • Knee length, worn directly over street clothes
  • Color: Royal Blue
  • Available in various sizes
  • ARC Rating: 5.7
  • HRC Category: 1
Compliance
Meets NFPA 2112 Standard on Flame Resistance Clothing for Protection of Industrial Personnel Against Short-Duration Thermal Exposure from Fire.
Specifications
Size Small
Material Nomex IIIA
Color Royal Blue