Learn More
Bulwark™ CP Chemical Splash Protection Lab Coats
Catalog No. 19823617 Shop All Red Kap Industries Products
This item is not returnable. View return policy
Catalog No. 19-823-617 Supplier Bulwark™ CP Supplier No. DISCONTINUED
This item is not returnable. View return policy
Bulwark™ CP Chemical Splash Protection Lab Coatc with Milliken ShieldCSR chemical splash protection finish that resists penetration and repels inadvertent liquid chemical splash.
  • Featuring Milliken ShieldCSR Chemical Splash protection finish resists penetration and repels inadvertent liquid chemical splash
  • Snap front closure for easy on/off
  • Lay-down collar
  • Splash resistant side pass-throughs for easy inner pocket access
  • Pockets include one welt chest pocket on left chest and two large lower front pockets with flaps
Specifications
Product Type Chemical Resistant Lab Coat
Gender Unisex
Color White
Disposable Reusable
Sterility Non-sterile
Resistance Chemicals