Learn More
Thermco Products Inc WORKSTATION THERMOMETER
Supplier:  Thermco Products Inc WS0013FC
Encompass_Preferred
Workstation Thermometer is designed to monitor the temperature of cells in the workstation reference card.The beaded probe is inserted into the card through the foil cover. Probe wire is 1mm in diameter and fits easily under the workstation cover.The thermometer has double Hook and Loop for attachment of the thermometer to the side of the workstation.The WS0013FC is certified at 37C and traceable to N.I.S.T. and ISO 17025/ILAC with a full calibration certificate.
Catalog No. NC0997838