Learn More
Worldwide Glass Resource 13-425 Polypropylene Screw Thread Closures
Shop All Worldwide Glass Resource ProductsDescription
- 13-425 screw thread finish
- Polypropylene construction
Specifications
Specifications
| Color | Black |
| Material | Polypropylene |
| Pre-slit | No |
| Closure Size | 13-425 |
| For Use With (Equipment) | Chromatography Vials |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.