Learn More
Eisco™ Worm Gear Drive Mechanical Demonstration Model
SCI_ED
Catalog No. S27675 Shop All Eisco Products
Change view
Click to view available options
Worm Gear Drive Mechanical Demonstration Model
Catalog No.
S27675 Worm Gear Drive Mechanical Demonstration Model
1 options
Catalog No. S27675 Supplier Eisco™ Supplier No. PH0366TM6
Add to Cart
Add to Cart
The Worm Gear Drive is an interactive visual demonstration of a common power transmission mechanism used in machinery, automotive systems, robotics, and conveyors.
  • Hand-cranked interactive model for visual learning.
  • Shows the fundamentals of how a worm gear works.
  • Dial to show rotation.
  • Open system is sandwiched by two clear plastic plates for unobstructed study.
  • Constructed with heavy duty metal and plastic.
  • Includes comprehensive instruction and experiment manual.
Specifications
Product Type Worm Gear Drive Mechanical Demonstration Model