Learn More
Grainger Threading Tool 1/2 Carbide
Supplier:  Grainger 16T193
Encompass_Preferred
The Micro 100 1/2" Shank Threading Tool is a high speed steel threading tool with a 1/2" shank diameter and an overall length of 5". It has a 60 degree threading point and can cut unified coarse, unified fine, and 8 pitch threads. The tool has an AlTiN nano coating for increased tool life and lubricity. It is made in the USA by Micro 100 and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, brass, bronze, plastics, and composites. The tool provides accurate threading with optimized chip control.
Catalog No. 50-276-8639
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.