Learn More
Grainger DOT Container Label 4 Label W PK500
Supplier:  Grainger 16U817
Encompass_Preferred
The Tripp Lite SMART1500LCDT UPS offers 1500VA/900W capacity with 8 outlets (5 battery backup & surge, 3 surge only). It provides battery backup and surge protection for electronics. The LCD shows diagnostics like runtime, load, and battery levels. It features automatic voltage regulation, tel/DSL protection, USB, and DB9 serial ports. The tower form factor includes software for shutdown, logging, and scheduling. ENERGY STAR certified.
Catalog No. 50-276-7262
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.