Learn More
Grainger Worm Gear Hose Clamp 5/8 W 4 3/4 ID PK10
Supplier:  Grainger 16P242
Encompass_Preferred
The Worm Gear Hose Clamp 301 Stainless Steel by Oetiker is a high-quality stainless steel worm gear clamp with a 301 stainless steel band. It has a band width of 7/16", a clamping range of 5/8" to 1-1/4", and a torque of 35 in-lbs. This heavy duty hose clamp provides a full 360 degree seal. It is made in the USA and conforms to SAE J1508 Type 1A. The clamp features a rolled edge for added strength.
Catalog No. 50-276-3949
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.